Learn More
Medchemexpress LLC Amthamine dihydrobromide | 142457-00-9 | MFCD00673891 | 99.6% | 319.07 g·mol⁻¹ | C6H13Br2N3S | 25 MG
SDP
Catalog No. 5000213239
Encompass_Preferred
Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a histamine receptor agonist active at H1-H4 receptors. It is supplied as a solid and in ready-to-use DMSO solution formats for pharmacological research, and includes storage recommendations for both solids and stock solutions.
Catalog No. 50-002-13239 Supplier Medchemexpress LLC Supplier No. HY10106325MG
Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a histamine receptor agonist active at H1-H4 receptors. It is supplied as a solid and in ready-to-use DMSO solution formats for pharmacological research, and includes storage recommendations for both solids and stock solutions.