Learn More
Medchemexpress LLC Amthamine dihydrobromide | 142457-00-9 | MFCD00673891 | 99.6% | 319.07 g·mol⁻¹ | C6H13Br2N3S | 5 MG
SDP
Catalog No. 5000212901
Encompass_Preferred
Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a selective histamine receptor agonist used in pharmacology and receptor research. It is supplied as a purified solid for use in in vitro and in vivo assays to probe histamine receptor function and signaling.
  • High reported purity: 99.6%.
  • Molecular weight 319.07 g·mol⁻¹.
  • Supplied as a solid dihydrobromide salt.
  • Suitable for histamine receptor pharmacology and assay work.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-12901 Supplier Medchemexpress LLC Supplier No. HY1010635MG
Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a selective histamine receptor agonist used in pharmacology and receptor research. It is supplied as a purified solid for use in in vitro and in vivo assays to probe histamine receptor function and signaling.
  • High reported purity: 99.6%.
  • Molecular weight 319.07 g·mol⁻¹.
  • Supplied as a solid dihydrobromide salt.
  • Suitable for histamine receptor pharmacology and assay work.
  • Available in small milligram pack sizes for research use.