Learn More
Medchemexpress LLC Amthamine dihydrobromide | 142457-00-9 | MFCD00673891 | 99.6% | 319.07 g/mol | C6H13Br2N3S | 50 MG
SDP
Catalog No. 5000213240
Encompass_Preferred
Amthamine dihydrobromide is a research-grade histamine receptor agonist provided as the dihydrobromide salt. It is used in pharmacology and toxicology research to study histamine receptor activation and downstream physiological effects in vitro and in vivo.
  • Selective histamine H2 receptor agonist.
  • High chemical purity suitable for research applications.
  • Available as a solid for flexible dosing and formulation.
  • Includes molecular and CAS identifiers for unambiguous compound identification.
Catalog No. 50-002-13240 Supplier Medchemexpress LLC Supplier No. HY10106350MG
Amthamine dihydrobromide is a research-grade histamine receptor agonist provided as the dihydrobromide salt. It is used in pharmacology and toxicology research to study histamine receptor activation and downstream physiological effects in vitro and in vivo.
  • Selective histamine H2 receptor agonist.
  • High chemical purity suitable for research applications.
  • Available as a solid for flexible dosing and formulation.
  • Includes molecular and CAS identifiers for unambiguous compound identification.