Learn More
Apexbio Technology LLC AMTHAMINE-DIHYDROBROMIDE-25MG
SDP
Catalog No. 5000569604
Encompass_Preferred
Amthamine dihydrobromide (CAS No 142457-00-9) is a potent histamine H2 receptor agonist primarily used in biomedical research Originating from studies on histaminergic pathways this compound selectively targets H2 receptors facilitating the study of gastric acid secretion and immune response modulation In vitro its activity is typically observed in the nanomolar range making it highly effective in precise mechanism studies Amthamine dihydrobromide is commonly applied in cell models to dissect receptor signaling and in animal studies to explore physiological responses linked to H2 receptor activation It is also valuable in drug screening processes for developing new therapeutics targeting histamine pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69604 Supplier Apexbio Technology LLC Supplier No. B640025
Amthamine dihydrobromide (CAS No 142457-00-9) is a potent histamine H2 receptor agonist primarily used in biomedical research Originating from studies on histaminergic pathways this compound selectively targets H2 receptors facilitating the study of gastric acid secretion and immune response modulation In vitro its activity is typically observed in the nanomolar range making it highly effective in precise mechanism studies Amthamine dihydrobromide is commonly applied in cell models to dissect receptor signaling and in animal studies to explore physiological responses linked to H2 receptor activation It is also valuable in drug screening processes for developing new therapeutics targeting histamine pathways Experimental concentrations typically depend on the specific research design