Learn More
Cayman Chemical AMT (hydrochloride), Purity: ≥98%, CAS: 21463-31-0, 50 mg
SDP
Catalog No. 502896231
Encompass_Preferred
AMT is a potent, relatively selective inhibitor of iNOS with a Ki of 4.2 nM in mouse macrophages. It inhibits other isoforms of NOS, but at higher concentrations; the IC50 values for rat nNOS and bovine eNOS are 34 and 150 nM, respectively.{1351}
Catalog No. 50-289-6231 Supplier Cayman Chemical Supplier No. 8101050 MG
AMT is a potent, relatively selective inhibitor of iNOS with a Ki of 4.2 nM in mouse macrophages. It inhibits other isoforms of NOS, but at higher concentrations; the IC50 values for rat nNOS and bovine eNOS are 34 and 150 nM, respectively.{1351}