Learn More
Medchemexpress LLC HY-10206 5mg Medchemexpress, Amuvatinib CAS:850879-09-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102065MG
Encompass_Preferred
Medchemexpress, HY-10206 5mg Amuvatinib CAS:850879-09-3 Amuvatinib (MP470) is an orally bioavailable multi-targeted tyrosine kinase inhibitor with potent activity against mutant c-Kit, PDGFRα, Flt3, c-Met and c-Ret. Amuvatinib (MP470) is also a DNA repair suppressor through suppression of DNA repair protein RAD51, thereby disrupting DNA damage repair[3]. Antineoplastic activity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7975