Learn More
Selleck Chemical LLC Amuvatinib (MP-470) S1244-2mg
SDP
Catalog No. 503111559
Encompass_Preferred
Amuvatinib (MP-470 HPK 56) is a potent and multi-targeted inhibitor of c-Kit PDGFR and Flt3 with IC50 of 10 nM 40 nM and 81 nM respectively Amuvatinib suppresses c-MET and c-RET Amuvatinib is also active as a DNA repair protein Rad51 inhibitor with antineoplastic activity Phase 2
Catalog No. 50-311-1559 Supplier Selleck Chemical LLC Supplier No. S12442MG
Amuvatinib (MP-470 HPK 56) is a potent and multi-targeted inhibitor of c-Kit PDGFR and Flt3 with IC50 of 10 nM 40 nM and 81 nM respectively Amuvatinib suppresses c-MET and c-RET Amuvatinib is also active as a DNA repair protein Rad51 inhibitor with antineoplastic activity Phase 2