Learn More
Selleck Chemical LLC Amuvatinib (MP-470) 50mg 850879-09-3 HPK 56
SDP
Catalog No. 508166
Encompass_Preferred
Amuvatinib (MP-470, HPK 56) is a potent and multi-targeted inhibitor of c-Kit, PDGFR? and Flt3 with IC50 of 10 nM, 40 nM and 81 nM, respectively. Amuvatinib suppresses c-MET and c-RET. Amuvatinib is also active as a DNA repair protein Rad51 inhibitor with antineoplastic activity. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-816-6 Supplier Selleck Chemical LLC Supplier No. S124450MG
Add to Cart
Add to Cart
Amuvatinib (MP-470, HPK 56) is a potent and multi-targeted inhibitor of c-Kit, PDGFR? and Flt3 with IC50 of 10 nM, 40 nM and 81 nM, respectively. Amuvatinib suppresses c-MET and c-RET. Amuvatinib is also active as a DNA repair protein Rad51 inhibitor with antineoplastic activity. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.