Learn More
Agilent BioTek AMX Automated Media Exchange
GSA_VAGREENER_CHOICE
Catalog No. BT1260041 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
MFXP2, MFXP2R
Catalog No. For Use With (Equipment)
BT1260041 MFXP2, MFXP2R
1 options
Catalog No. BT1260041 Supplier Agilent BioTek Supplier No. 1260041
Add to Cart
Add to Cart
Request a Quote
AMX Automated Media Exchange module protects 3D cell structures (e.g. spheroids, tumoroids), and suspension cells
  • AMX Automated Media Exchange module protects 3D cell structures (e.g. spheroids, tumoroids), and suspension cells
  • For use with Agilent BioTek Microplate readers and imagers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE and TUV marked. RoHS compliant.
Description AMX Automated Media Exhange module
For Use With (Application) Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization
For Use With (Equipment) MFXP2, MFXP2R
Includes AMX Autmated Media Exchange module, second peri pump, AMX dispense and aspirate manifolds