Learn More
Medchemexpress LLC Amxi-5001 | 2170491-77-5 | MFCD34676674 | 98.3% | 457.46 | C25H20FN5O3 | 5 MG
SDP
Catalog No. 5000221098
Encompass_Preferred
AMXI-5001 is a small-molecule research compound described as a potent, orally active dual PARP1/2 and microtubule polymerization inhibitor with selective antitumor cytotoxicity in multiple human cancer cell lines; it has demonstrated tumor regression in preclinical studies.
  • Potent dual PARP1/2 and microtubule polymerization inhibition.
  • Selective antitumor cytotoxicity across various human cancer cell lines.
  • High purity suitable for research applications.
  • Soluble in DMSO at 50 mg/mL with warming and sonication recommended.
  • Stable as a powder when stored at -20°C for long-term storage.
Catalog No. 50-002-21098 Supplier Medchemexpress LLC Supplier No. HY1457345MG
AMXI-5001 is a small-molecule research compound described as a potent, orally active dual PARP1/2 and microtubule polymerization inhibitor with selective antitumor cytotoxicity in multiple human cancer cell lines; it has demonstrated tumor regression in preclinical studies.
  • Potent dual PARP1/2 and microtubule polymerization inhibition.
  • Selective antitumor cytotoxicity across various human cancer cell lines.
  • High purity suitable for research applications.
  • Soluble in DMSO at 50 mg/mL with warming and sonication recommended.
  • Stable as a powder when stored at -20°C for long-term storage.