Learn More
Medchemexpress LLC AMXI-5001 hydrochloride | 00-00-0 | 98.1% | C25H21ClFN5O3 | 10MG
SDP
Catalog No. 5000258751
Encompass_Preferred
AMXI-5001 hydrochloride is a research-grade small molecule described as a dual inhibitor of PARP1/2 and microtubule polymerization. It shows potent cellular activity against PARP and tubulin targets, with reported favorable metabolic stability and oral bioavailability, and is intended for preclinical anticancer research.
  • Dual mechanism: inhibits PARP1/2 and tubulin polymerization.
  • Potent PARP inhibition with low nanomolar IC50s and tubulin inhibition in the micromolar range.
  • Orally bioavailable with favorable pharmacokinetic properties.
  • Suitable for in vitro and in vivo anticancer studies.
  • High purity for research applications.
Catalog No. 50-002-58751 Supplier Medchemexpress LLC Supplier No. HY145734A10MG
AMXI-5001 hydrochloride is a research-grade small molecule described as a dual inhibitor of PARP1/2 and microtubule polymerization. It shows potent cellular activity against PARP and tubulin targets, with reported favorable metabolic stability and oral bioavailability, and is intended for preclinical anticancer research.
  • Dual mechanism: inhibits PARP1/2 and tubulin polymerization.
  • Potent PARP inhibition with low nanomolar IC50s and tubulin inhibition in the micromolar range.
  • Orally bioavailable with favorable pharmacokinetic properties.
  • Suitable for in vitro and in vivo anticancer studies.
  • High purity for research applications.