Learn More
Medchemexpress LLC AMXI-5001 hydrochloride | 2170491-77-5 | 98.1% | C25H21ClFN5O3 | 25 MG
SDP
Catalog No. 5000353125
Encompass_Preferred
AMXI-5001 hydrochloride is a potent, orally active, dual PARP1/2 and microtubule polymerization inhibitor. It shows selective antitumor cytotoxicity against various human cancer cells with significantly lower IC50s compared to existing clinical PARP1/2 inhibitors. This compound has been observed to induce complete regression of established tumors, even exceedingly large ones.
  • Potent, orally active, dual PARP1/2 and microtubule polymerization inhibitor
  • Shows selective antitumor cytotoxicity against various human cancer cells
  • Significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors, even exceedingly large ones
Catalog No. 50-003-53125 Supplier Medchemexpress LLC Supplier No. 25MGHY145734A
AMXI-5001 hydrochloride is a potent, orally active, dual PARP1/2 and microtubule polymerization inhibitor. It shows selective antitumor cytotoxicity against various human cancer cells with significantly lower IC50s compared to existing clinical PARP1/2 inhibitors. This compound has been observed to induce complete regression of established tumors, even exceedingly large ones.
  • Potent, orally active, dual PARP1/2 and microtubule polymerization inhibitor
  • Shows selective antitumor cytotoxicity against various human cancer cells
  • Significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors, even exceedingly large ones