Learn More
Medchemexpress LLC AMXI-5001 hydrochloride | 98.1% | C25H21ClFN5O3 | 1 ML
SDP
Catalog No. 5000355490
Encompass_Preferred
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It shows selective antitumor cytotoxicity across a wide range of human cancer cells with significantly lower IC50s compared to existing clinical PARP1/2 inhibitors, and can induce complete regression of established tumors, including very large ones.
  • Potent and orally active
  • Dual PARP1/2 and microtubule polymerization inhibitor
  • Selective antitumor cytotoxicity across a wide range of human cancer cells
  • Significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors
Catalog No. 50-003-55490 Supplier Medchemexpress LLC Supplier No. HY145734A10MM/1ML
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It shows selective antitumor cytotoxicity across a wide range of human cancer cells with significantly lower IC50s compared to existing clinical PARP1/2 inhibitors, and can induce complete regression of established tumors, including very large ones.
  • Potent and orally active
  • Dual PARP1/2 and microtubule polymerization inhibitor
  • Selective antitumor cytotoxicity across a wide range of human cancer cells
  • Significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors