Learn More
Medchemexpress LLC AMXI-5001 hydrochloride | 98.1% | C25H21ClFN5O3 | 100 MG
SDP
Catalog No. 5000354160
Encompass_Preferred
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It exhibits selective antitumor cytotoxicity across a wide variety of human cancer cells with much lower IC50s than existing clinical PARP1/2 inhibitors. This compound induces complete regression of established tumors, even very large ones. It is intended for research use only and not sold to patients.
  • Potent and orally active
  • Functions as a dual PARP1/2 and microtubule polymerization inhibitor
  • Demonstrates selective antitumor cytotoxicity against various human cancer cells
  • Has significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors
  • For research use only
Catalog No. 50-003-54160 Supplier Medchemexpress LLC Supplier No. HY145734A100MG
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It exhibits selective antitumor cytotoxicity across a wide variety of human cancer cells with much lower IC50s than existing clinical PARP1/2 inhibitors. This compound induces complete regression of established tumors, even very large ones. It is intended for research use only and not sold to patients.
  • Potent and orally active
  • Functions as a dual PARP1/2 and microtubule polymerization inhibitor
  • Demonstrates selective antitumor cytotoxicity against various human cancer cells
  • Has significantly lower IC50s compared to existing clinical PARP1/2 inhibitors
  • Induces complete regression of established tumors
  • For research use only