Learn More
Medchemexpress LLC Amxi-5001 hydrochloride | 98.1% | C25H21ClFN5O3 | 50 MG
SDP
Catalog No. 5000355996
Encompass_Preferred
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It exhibits selective antitumor cytotoxicity across a wide variety of human cancer cells, with much lower IC50s than existing clinical PARP1/2 inhibitors. This compound induces complete regression of established tumors, including exceedingly large tumors. It is intended for research use only.
  • Potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor
  • Exhibits selective antitumor cytotoxicity across various human cancer cells
  • Induces complete regression of established tumors, including large ones
Catalog No. 50-003-55996 Supplier Medchemexpress LLC Supplier No. HY14573450MG
AMXI-5001 hydrochloride is a potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor. It exhibits selective antitumor cytotoxicity across a wide variety of human cancer cells, with much lower IC50s than existing clinical PARP1/2 inhibitors. This compound induces complete regression of established tumors, including exceedingly large tumors. It is intended for research use only.
  • Potent, orally active, and dual PARP1/2 and microtubule polymerization inhibitor
  • Exhibits selective antitumor cytotoxicity across various human cancer cells
  • Induces complete regression of established tumors, including large ones