Learn More
Medchemexpress LLC HY-124617A 10mg , AMXT-1501 tetrahydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124617A10MG
Encompass_Preferred
HY-124617A 10mg AMXT-1501 tetrahydrochloride CAS: AMXT-1501 tetrahydrochloride is an orally active polyamine transport inhibitor. AMXT1501 blocks tumor growth in immunocompetent mice but not in athymic nude mice lacking T cells. Combination of DFMO and AMXT‐1501 induces caspase‐3 mediated apoptosis in NB cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0630