Learn More
Medchemexpress LLC HY-124617A 5mg Medchemexpress, AMXT-1501 tetrahydrochloride CAS: Purity:>98%
SDP
Catalog No. 501969670
Encompass_Preferred
Medchemexpress, HY-124617A 5mg AMXT-1501 tetraHCl CAS: AMXT-1501 tetrahydrochloride is a polyamine transport inhibitor. AMXT1501 blocks tumor growth in immunocompetent mice but not in athymic nude mice lacking T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9670 Supplier Medchemexpress LLC Supplier No. HY124617A5MG
Medchemexpress, HY-124617A 5mg AMXT-1501 tetraHCl CAS: AMXT-1501 tetrahydrochloride is a polyamine transport inhibitor. AMXT1501 blocks tumor growth in immunocompetent mice but not in athymic nude mice lacking T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.