Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amxt-1501 tetrahydrochloride | 2444815-84-1 | 98.0% | 714.77 g/mol | C32H72Cl4N6O2 | 50MG
SDP

Catalog No. 5000236145
Encompass_Preferred

AMXT-1501 tetrahydrochloride is the tetrahydrochloride salt of AMXT-1501, a research-stage inhibitor of the polyamine transport system used in preclinical oncology studies. Supplied for research use, it is characterized by a molecular weight of 714.77 g/mol, molecular formula C32H72Cl4N6O2, and NMR purity of 98.0%.

  • Research-grade polyamine transport inhibitor
  • High NMR purity (98.0%)
  • Molecular weight 714.77 g/mol
  • Chemical formula C32H72Cl4N6O2
  • Provided as a tetrahydrochloride salt
  • Intended for preclinical research use only

Catalog No. 50-002-36145 Supplier Medchemexpress LLC Supplier No. HY124617A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMXT-1501 tetrahydrochloride is the tetrahydrochloride salt of AMXT-1501, a research-stage inhibitor of the polyamine transport system used in preclinical oncology studies. Supplied for research use, it is characterized by a molecular weight of 714.77 g/mol, molecular formula C32H72Cl4N6O2, and NMR purity of 98.0%.

  • Research-grade polyamine transport inhibitor
  • High NMR purity (98.0%)
  • Molecular weight 714.77 g/mol
  • Chemical formula C32H72Cl4N6O2
  • Provided as a tetrahydrochloride salt
  • Intended for preclinical research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.