Learn More
Medchemexpress LLC AMY-101 | 1427001-89-5 | 99.86% | 1789.09 | 10 MG
SDP
Catalog No. 5000423681
Encompass_Preferred
AMY-101 (Cp40) is a peptidic inhibitor of the central complement component C3 with a KD of 0.5 nM. It has been shown to inhibit naturally occurring periodontitis in non-human primates (NHPs) and exhibits favorable anti-inflammatory activity in models of COVID-19 severe pneumonia with systemic hyper-inflammation.
  • Inhibits central complement component C3
  • Improves periodontal condition in NHPs
  • Induces a long-lasting anti-inflammatory effect
  • Attenuates fibrosis and infiltration of inflammatory cells in UUO-induced renal fibrosis
Catalog No. 50-004-23681 Supplier Medchemexpress LLC Supplier No. HYP171710MG
AMY-101 (Cp40) is a peptidic inhibitor of the central complement component C3 with a KD of 0.5 nM. It has been shown to inhibit naturally occurring periodontitis in non-human primates (NHPs) and exhibits favorable anti-inflammatory activity in models of COVID-19 severe pneumonia with systemic hyper-inflammation.
  • Inhibits central complement component C3
  • Improves periodontal condition in NHPs
  • Induces a long-lasting anti-inflammatory effect
  • Attenuates fibrosis and infiltration of inflammatory cells in UUO-induced renal fibrosis