Learn More
Medchemexpress LLC HY-P1717B 5mg , AMY-101 (acetate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1717B5MG
Encompass_Preferred
HY-P1717B 5mg AMY-101 (acetate) CAS: AMY-101 acetate (Cp40 acetate), a peptidic inhibitor of the central complement component C3 (KD = 0.5 nM), inhibits naturally occurring periodontitis in non-human primates (NHPs). AMY-101 acetate (Cp40 acetate) exhibits a favorable anti-inflammatory activity in models with COVID-19 severe pneumonia with systemic hyper inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1210