Learn More
Medchemexpress LLC Amy-101 (Cp40) | 1427001-89-5 | 99.9% | 1789.09 | 5 MG
SDP
Catalog No. 5000423905
Encompass_Preferred
AMY-101 (Cp40) is a peptidic inhibitor of the central complement component C3 with a KD of 0.5 nM. It has been shown to inhibit naturally occurring periodontitis in non-human primates (NHPs) and demonstrates favorable anti-inflammatory activity in models of COVID-19 severe pneumonia with systemic hyper inflammation.
  • Inhibits naturally occurring periodontitis
  • Exhibits anti-inflammatory activity in models with COVID-19 severe pneumonia
  • Can improve periodontal condition of NHPs
  • Induces a long-lasting anti-inflammatory effect
  • Attenuates fibrosis and infiltration of inflammatory cells in UUO-induced renal fibrosis
Catalog No. 50-004-23905 Supplier Medchemexpress LLC Supplier No. HYP17175MG
AMY-101 (Cp40) is a peptidic inhibitor of the central complement component C3 with a KD of 0.5 nM. It has been shown to inhibit naturally occurring periodontitis in non-human primates (NHPs) and demonstrates favorable anti-inflammatory activity in models of COVID-19 severe pneumonia with systemic hyper inflammation.
  • Inhibits naturally occurring periodontitis
  • Exhibits anti-inflammatory activity in models with COVID-19 severe pneumonia
  • Can improve periodontal condition of NHPs
  • Induces a long-lasting anti-inflammatory effect
  • Attenuates fibrosis and infiltration of inflammatory cells in UUO-induced renal fibrosis