Learn More
Medchemexpress LLC HY-P1717A 1mg Medchemexpress, AMY-101 (TFA) CAS:1789738-04-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1717A1MG
Encompass_Preferred
Medchemexpress, HY-P1717A 1mg AMY-101 (TFA) CAS:1789738-04-0 AMY-101 TFA (Cp40 TFA), a peptidic inhibitor of the central complement component C3 (KD = 0.5 nM), inhibits naturally occurring periodontitis in non-human primates (NHPs). AMY-101 (Cp40) exhibits a favorable anti-inflammatory activity in models with COVID-19 severe pneumonia with systemic hyper inflammation[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1697