Learn More
Aviva Systems Biology Corp AMY2B Antibody - C-terminal region (ARP61947_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61947_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMY2B. It was validated on Western Blot. . Alternate names: AMY2ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-517