Learn More
Cayman Chemical AmycolatopsIn B 100ug
SDP
Catalog No. 502991258
Encompass_Preferred
A bacterial metabolite; cytotoxic to NCI H460 lung and SW620 colon cancer cells (IC50s = 0.28 and 0.14 µM, respectively)
Catalog No. 50-299-1258 Supplier Cayman Chemical Supplier No. 31677100 UG
A bacterial metabolite; cytotoxic to NCI H460 lung and SW620 colon cancer cells (IC50s = 0.28 and 0.14 µM, respectively)