Learn More
Alkali Scientific Amygdalin [D Mandelonitrile b gentiobioside] [Laetrile], 5 G
SDP
Catalog No. 5000661091
Encompass_Preferred
Amygdalin is a plant-derived glycoside used in biochemical research, especially in studies related to cyanogenic compounds. This 5g laboratory-grade sample is used to investigate the metabolic breakdown of glycosides and the potential release of cyanide. Amygdalin is also studied for its possible therapeutic properties, particularly in cancer research, where it is believed to release cyanide in a controlled manner that could target cancer cells. The 5g bottle is ideal for small-scale experiments and research focused on enzymatic reactions, plant defense mechanisms, and bioassays related to cyanogenic compounds.
Catalog No. 50-006-61091 Supplier Alkali Scientific Supplier No. AMY003CP
Amygdalin is a plant-derived glycoside used in biochemical research, especially in studies related to cyanogenic compounds. This 5g laboratory-grade sample is used to investigate the metabolic breakdown of glycosides and the potential release of cyanide. Amygdalin is also studied for its possible therapeutic properties, particularly in cancer research, where it is believed to release cyanide in a controlled manner that could target cancer cells. The 5g bottle is ideal for small-scale experiments and research focused on enzymatic reactions, plant defense mechanisms, and bioassays related to cyanogenic compounds.