Learn More
Cayman Chemical Amygdalin, Purity: ≥98%, CAS: 29883-15-6, 25 g
SDP
Catalog No. 502934174
Encompass_Preferred
A glycoside with diverse biological activities; induces cell cycle arrest at the G0/G1 phase, decreases cyclin A and Cdk2 levels, and inhibits cell growth in UMUC-3, RT112, and TCCSUP bladder cancer cells from 1.25-10 mg/ml; reduces the number of primary microtubules and microvessels in aortic rings isolated from rats with streptozotocin-induced diabetes at 3 mg/kg; reduces triglyceride, total cholesterol, and LDL levels and aortic sinus plaque area in an LDLR-/- mouse model of atherosclerosis; reduces production of TNF-α, IL-1β, and IL-6 as well as neutrophil and macrophage infiltration in BALF in a mouse model of LPS-induced acute lung injury
Catalog No. 50-293-4174 Supplier Cayman Chemical Supplier No. 2666825 G
A glycoside with diverse biological activities; induces cell cycle arrest at the G0/G1 phase, decreases cyclin A and Cdk2 levels, and inhibits cell growth in UMUC-3, RT112, and TCCSUP bladder cancer cells from 1.25-10 mg/ml; reduces the number of primary microtubules and microvessels in aortic rings isolated from rats with streptozotocin-induced diabetes at 3 mg/kg; reduces triglyceride, total cholesterol, and LDL levels and aortic sinus plaque area in an LDLR-/- mouse model of atherosclerosis; reduces production of TNF-α, IL-1β, and IL-6 as well as neutrophil and macrophage infiltration in BALF in a mouse model of LPS-induced acute lung injury