Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate (Laboratory), Fisher Chemical™

Catalog No. A7184 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A718-4 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A718-4

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C

Specifications

Melting Point -70.8°C
Color Colorless
Density 0.870g/cm³
Boiling Point 149°C
Flash Point 40°C
Packaging Poly Bottle
Linear Formula CH3(CH2)4OC(O)CH3
Quantity 4 L
UN Number 1104
Formula Weight 130.19
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Amyl Acetate
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.