Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Amyl Acetate Reagent A.C.S. 1 QT
SDP

Catalog No. 50980181
Encompass_Preferred

(Molybdic acid, ammonium salt)
CAS #628-63-7 CH₃COOC₅H₁₁ F.W. 130.18
Boiling point: 140.0°C-150.0°C @ 5 mmHg

Catalog No. 50-980-181 Supplier Electron Microscopy Sciences Supplier No. 10810
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Molybdic acid, ammonium salt)
CAS #628-63-7 CH₃COOC₅H₁₁ F.W. 130.18
Boiling point: 140.0°C-150.0°C @ 5 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.