Learn More
Sigma Aldrich Fine Chemicals Biosciences Amyl butyrate 98 FG
Catalog No. 502907346
Encompass
Amyl butyrate is an odor-active volatile organic compound that has been identified as one of the main flavor constituents in banana fruit apricot and apples. It is used as food flavoring agent in chewing gums candies and baked goods.
Catalog No. 50-290-7346 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W20591520KGK
Amyl butyrate is an odor-active volatile organic compound that has been identified as one of the main flavor constituents in banana fruit apricot and apples. It is used as food flavoring agent in chewing gums candies and baked goods.