Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Butyrate, MP Biomedicals

Catalog No. ICN213130 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213130 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213130 Supplier MP Biomedicals, Inc Supplier No. 0521313080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 540-18-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00048850
InChI Key CFNJLPHOBMVMNS-UHFFFAOYSA-N
Synonym pentyl butyrate, amyl butyrate, n-amyl butyrate, butanoic acid, pentyl ester, amyl butanoate, 1-pentyl butyrate, n-pentyl butanoate, n-pentyl n-butyrate, n-pentyl butyrate, butyric acid, pentyl ester
PubChem CID 10890
ChEBI CHEBI:87684
IUPAC Name pentyl butanoate
SMILES CCCCCOC(=O)CCC

Specifications

Quantity 100 g
Formula Weight 158.24
Chemical Name or Material Amyl Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.