Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. B075525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B075525ML 25 mL
B0755500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B075525ML Supplier TCI America Supplier No. B075525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-18-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00048850
InChI Key CFNJLPHOBMVMNS-UHFFFAOYSA-N
Synonym pentyl butyrate, amyl butyrate, n-amyl butyrate, butanoic acid, pentyl ester, amyl butanoate, 1-pentyl butyrate, n-pentyl butanoate, n-pentyl n-butyrate, n-pentyl butyrate, butyric acid, pentyl ester
PubChem CID 10890
ChEBI CHEBI:87684
IUPAC Name pentyl butanoate
SMILES CCCCCOC(=O)CCC

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 mL
UN Number 2620
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET5956000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.