Learn More
YMC America, Inc Amyl-C Neo guard 10um 10x4mm
SDP
Catalog No. 5000881538
Encompass
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes
Catalog No. 50-008-81538 Supplier YMC America, Inc Supplier No. KBN99S110104GC
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes