Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Laurate 97.0+%, TCI America™
SDP

Catalog No. L001225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001225ML 25 mL
L0012500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L001225ML Supplier TCI America Supplier No. L001225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5350-03-8
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.46
MDL Number MFCD00059218
InChI Key PQWBDPUBNMEITD-UHFFFAOYSA-N
Synonym Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester
PubChem CID 62571
IUPAC Name pentyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCCCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 270.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Laurate
EINECSNumber (2)-0798
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.