Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl n-Octanoate 98.0+%, TCI America™
SDP

Catalog No. O002825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002825ML 25 mL
O0028500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O002825ML Supplier TCI America Supplier No. O002825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 638-25-5
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00048919
InChI Key GJWGZSBNFSBUPX-UHFFFAOYSA-N
Synonym amyl octanoate, amyl caprylate, octanoic acid, pentyl ester, amyl octoate, amyl octylate, pentyl octylate, amyl n-octanoate, n-caprylic acid n-amyl ester, n-amyl n-octanoate, unii-1kwp15429r
PubChem CID 61185
IUPAC Name pentyl octanoate
SMILES CCCCCCCC(=O)OCCCCC

Specifications

Color Yellow
Boiling Point 260°C
Quantity 25 mL
Formula Weight 214.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl n-Octanoate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.