Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Valerate, MP Biomedicals

Catalog No. ICN213052 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213052 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213052 Supplier MP Biomedicals, Inc Supplier No. 0521305280

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2173-56-0
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00042904
InChI Key FGPPDYNPZTUNIU-UHFFFAOYSA-N
Synonym pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester
PubChem CID 62433
IUPAC Name pentyl pentanoate
SMILES CCCCCOC(=O)CCCC

Specifications

Quantity 100 g
Formula Weight 172.27
Chemical Name or Material Amyl Valerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.