Learn More
Medchemexpress LLC HY-B2193 1g Medchemexpress, α-Amylase CAS:9000-90-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB21931G
Encompass_Preferred
Medchemexpress, HY-B2193 1g α-Amylase CAS:9000-90-2 α-Amylase is a hydrolase enzyme that catalyses the hydrolysis of internal α-1, 4-glycosidic linkages in starch to yield products like glucose and maltose. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3104