Learn More
Medchemexpress LLC Amylase | 9000-92-4 | 1 G
SDP
Catalog No. 5000411608
Encompass_Preferred
Amylase is an enzyme produced by the pancreas and salivary glands that catalyzes the hydrolysis of starch into sugars. Amylase is broadly classified into α, β, and γ subtypes. It is for research use only. A mixture of glycosylated amylase and proline-rich proteins or fractions containing mostly non-glycosylated amylase predominantly promotes adhesion of *S. gordonii G9B*. Fractions containing amylase do not promote adhesion of *S. sanguis 10556* to HAP.
  • Catalyzes the hydrolysis of starch into sugars
  • Available in α, β, and γ subtypes
  • Suitable for research applications
  • Offered in various quantities
  • Includes data sheet, SDS, COA, and handling instructions
  • Appears as a solid, with a color ranging from light yellow to khaki
  • Can be sourced from plants, animals, and microorganisms
Catalog No. 50-004-11608 Supplier Medchemexpress LLC Supplier No. HYB21921G
Amylase is an enzyme produced by the pancreas and salivary glands that catalyzes the hydrolysis of starch into sugars. Amylase is broadly classified into α, β, and γ subtypes. It is for research use only. A mixture of glycosylated amylase and proline-rich proteins or fractions containing mostly non-glycosylated amylase predominantly promotes adhesion of *S. gordonii G9B*. Fractions containing amylase do not promote adhesion of *S. sanguis 10556* to HAP.
  • Catalyzes the hydrolysis of starch into sugars
  • Available in α, β, and γ subtypes
  • Suitable for research applications
  • Offered in various quantities
  • Includes data sheet, SDS, COA, and handling instructions
  • Appears as a solid, with a color ranging from light yellow to khaki
  • Can be sourced from plants, animals, and microorganisms