Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Amylase from Aspergillus oryzae | 9001-19-8 | 10 G
SDP

Catalog No. 5000400379
Encompass_Preferred

α-Amylase from Aspergillus oryzae is an amylase utilized for various biochemical studies. This enzyme is provided as a white to off-white solid and is intended for research use only.

  • EC number: 3.2.1.1
  • Enzyme activity of at least 100 U/mg solid
  • One unit defined as the amount of enzyme required to liquefy 1 mg of soluble starch into dextrin in 1 minute at 70°C and pH 6.0
  • Soluble in water or according to specific experimental reference literature
  • Store under recommended conditions as per Certificate of Analysis

Catalog No. 50-004-00379 Supplier Medchemexpress LLC Supplier No. HYP276210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Amylase from Aspergillus oryzae is an amylase utilized for various biochemical studies. This enzyme is provided as a white to off-white solid and is intended for research use only.

  • EC number: 3.2.1.1
  • Enzyme activity of at least 100 U/mg solid
  • One unit defined as the amount of enzyme required to liquefy 1 mg of soluble starch into dextrin in 1 minute at 70°C and pH 6.0
  • Soluble in water or according to specific experimental reference literature
  • Store under recommended conditions as per Certificate of Analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.