Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Amylase from Aspergillus oryzae | 9001-19-8 | 100 G
SDP

Catalog No. 5000398678
Encompass_Preferred

α-Amylase from Aspergillus oryzae is an amylase suitable for various biochemical studies. It is intended for research use only.

  • CAS number 9001-19-8
  • EC number 3.2.1.1
  • Solid, white to off-white appearance
  • Enzyme activity of ≥100 U/mg solid
  • Targets amylases
  • Part of the metabolic enzyme/protease pathway
  • Store product under recommended conditions in Certificate of Analysis
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Catalog No. 50-003-98678 Supplier Medchemexpress LLC Supplier No. HYP2762100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Amylase from Aspergillus oryzae is an amylase suitable for various biochemical studies. It is intended for research use only.

  • CAS number 9001-19-8
  • EC number 3.2.1.1
  • Solid, white to off-white appearance
  • Enzyme activity of ≥100 U/mg solid
  • Targets amylases
  • Part of the metabolic enzyme/protease pathway
  • Store product under recommended conditions in Certificate of Analysis
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.