Learn More
Medchemexpress LLC α-Amylase from Aspergillus oryzae | 9001-19-8 | ≥100 U/mg solid | 25 G
SDP
Catalog No. 5000396233
Encompass_Preferred
Alpha-Amylase from Aspergillus oryzae is an amylase primarily utilized in various biochemical studies. This enzyme functions to liquefy soluble starch into dextrin, with a defined activity indicating its potency and effectiveness for experimental applications.
  • Can be used for various biochemical studies.
  • Enzyme activity is equal to or greater than 100 U/mg solid.
  • One unit liquefies 1mg of soluble starch into dextrin in one minute at 70°C and pH 6.0.
  • Appearance is solid, white to off-white.
  • Ships at room temperature in continental US.
  • Store under recommended conditions in the Certificate of Analysis.
Catalog No. 50-003-96233 Supplier Medchemexpress LLC Supplier No. 25GHYP2762
Alpha-Amylase from Aspergillus oryzae is an amylase primarily utilized in various biochemical studies. This enzyme functions to liquefy soluble starch into dextrin, with a defined activity indicating its potency and effectiveness for experimental applications.
  • Can be used for various biochemical studies.
  • Enzyme activity is equal to or greater than 100 U/mg solid.
  • One unit liquefies 1mg of soluble starch into dextrin in one minute at 70°C and pH 6.0.
  • Appearance is solid, white to off-white.
  • Ships at room temperature in continental US.
  • Store under recommended conditions in the Certificate of Analysis.