Learn More
Medchemexpress LLC α-Amylase from Aspergillus oryzae | 9001-19-8 | ≥100 U/mg solid | 5 G
SDP
Catalog No. 5000397372
Encompass_Preferred
Alpha-Amylase from Aspergillus oryzae is an amylase commonly used for various biochemical studies. This enzyme appears as a white to off-white solid and can be dissolved in water or other specific experimental reference literature as needed.
  • Used for various biochemical studies
  • Classified as an amylase enzyme
  • Appears as a white to off-white solid
  • Has an EC number of 3.2.1.1
  • Exhibits enzyme activity of ≥100 U/mg solid
  • Can be dissolved in water or according to specific experimental reference literature
Catalog No. 50-003-97372 Supplier Medchemexpress LLC Supplier No. HYP27625G
Alpha-Amylase from Aspergillus oryzae is an amylase commonly used for various biochemical studies. This enzyme appears as a white to off-white solid and can be dissolved in water or other specific experimental reference literature as needed.
  • Used for various biochemical studies
  • Classified as an amylase enzyme
  • Appears as a white to off-white solid
  • Has an EC number of 3.2.1.1
  • Exhibits enzyme activity of ≥100 U/mg solid
  • Can be dissolved in water or according to specific experimental reference literature