Learn More
Medchemexpress LLC α-Amylase from Aspergillus oryzae | 9001-19-8 | 50 G
SDP
Catalog No. 5000397456
Encompass_Preferred
α-Amylase from Aspergillus oryzae is an amylase used in various biochemical studies. This white to off-white solid is stable under recommended storage conditions and is intended for research use only by qualified personnel.
  • Used for biochemical studies
  • Laboratory chemical
  • White to off-white solid
  • Stable under recommended storage conditions
  • Enzyme activity of ≥100 U/mg solid
  • Soluble in water
Catalog No. 50-003-97456 Supplier Medchemexpress LLC Supplier No. HYP276250G
α-Amylase from Aspergillus oryzae is an amylase used in various biochemical studies. This white to off-white solid is stable under recommended storage conditions and is intended for research use only by qualified personnel.
  • Used for biochemical studies
  • Laboratory chemical
  • White to off-white solid
  • Stable under recommended storage conditions
  • Enzyme activity of ≥100 U/mg solid
  • Soluble in water