Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Amylase from Bacillus licheniformis, MilliporeSigma™ Supelco™

Catalog No. 111015766 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5766 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5766 Supplier MilliporeSigma Supelco Supplier No. A4582.5ML
Add to Cart
Add to Cart

α-Amylase belongs to glycosyl hydrolase family 13.

Featured Industries: Food and Beverages and General Analytical
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00081319
Synonym 1,4-alpha-D-Glucan-glucanohydrolase

Specifications

CAS 9000-85-5
Quantity 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.