Learn More
Medchemexpress LLC α-Amylase-IN-1 | 148404-10-8 | 98.3% | 310.35 | 1 MG
SDP
Catalog No. 5000361188
Encompass_Preferred
α-Amylase-IN-1 is an α-Amylase inhibitor with an IC50 value of 0.5509 μM. It also possesses antioxidant activity with an IC50 value of 53.49 μM for scavenging DPPH free radicals, making it suitable for the study of diabetes and oxidative stress-related diseases.
  • Potent α-Amylase inhibitor
  • Exhibits antioxidant activity
  • Useful in diabetes and oxidative stress-related diseases research
  • White to off-white solid
  • Soluble in DMSO
Catalog No. 50-003-61188 Supplier Medchemexpress LLC Supplier No. HY1493131MG
α-Amylase-IN-1 is an α-Amylase inhibitor with an IC50 value of 0.5509 μM. It also possesses antioxidant activity with an IC50 value of 53.49 μM for scavenging DPPH free radicals, making it suitable for the study of diabetes and oxidative stress-related diseases.
  • Potent α-Amylase inhibitor
  • Exhibits antioxidant activity
  • Useful in diabetes and oxidative stress-related diseases research
  • White to off-white solid
  • Soluble in DMSO