Learn More
Medchemexpress LLC Α-Amylase-IN-1 | 148404-10-8 | 98.3% | 310.35 | 10 MG
SDP
Catalog No. 5000365314
Encompass_Preferred
Α-Amylase-IN-1 (Compound 11) is an α-Amylase inhibitor with an IC50 value of 0.5509 μM. It also demonstrates antioxidant activity, with an IC50 value of 53.49 μM for scavenging DPPH free radicals. This compound can be utilized in the study of diabetes and oxidative stress-related diseases.
  • Functions as an α-Amylase inhibitor
  • Demonstrates antioxidant activity
  • Useful for studying diabetes
  • Applicable in research on oxidative stress-related diseases
Catalog No. 50-003-65314 Supplier Medchemexpress LLC Supplier No. 10MGHY149313
Α-Amylase-IN-1 (Compound 11) is an α-Amylase inhibitor with an IC50 value of 0.5509 μM. It also demonstrates antioxidant activity, with an IC50 value of 53.49 μM for scavenging DPPH free radicals. This compound can be utilized in the study of diabetes and oxidative stress-related diseases.
  • Functions as an α-Amylase inhibitor
  • Demonstrates antioxidant activity
  • Useful for studying diabetes
  • Applicable in research on oxidative stress-related diseases