Learn More
Medchemexpress LLC α-Amylase-IN-1 | 148404-10-8 | 98.3% | 310.35 | 5 MG
SDP
Catalog No. 5000360867
Encompass_Preferred
α-Amylase-IN-1 is a potent α-amylase inhibitor that also exhibits antioxidant activity. This compound is valuable for research into diabetes and various oxidative stress-related diseases.
  • Potent α-amylase inhibitor
  • Possesses antioxidant activity
  • Used in the study of diabetes
  • Used in the study of oxidative stress-related diseases
  • For research use only
  • Stable under recommended storage conditions
Catalog No. 50-003-60867 Supplier Medchemexpress LLC Supplier No. HY1493135MG
α-Amylase-IN-1 is a potent α-amylase inhibitor that also exhibits antioxidant activity. This compound is valuable for research into diabetes and various oxidative stress-related diseases.
  • Potent α-amylase inhibitor
  • Possesses antioxidant activity
  • Used in the study of diabetes
  • Used in the study of oxidative stress-related diseases
  • For research use only
  • Stable under recommended storage conditions