Learn More
Medchemexpress LLC α-Amylase-IN-3 | 93944-58-2 | 99.5% | 267.24 | 1 ML
SDP
Catalog No. 5000360827
Encompass_Preferred
α-Amylase-IN-3 (Compound 4) is an inhibitor for α-Amylase, AChE, and BChE, with respective IC50 values of 18.04 μM, 21.04 μM, and 22.2 μM. It demonstrates antioxidant activity and is utilized in research concerning diabetes and oxidative stress-associated diseases. The product is supplied as a white to off-white solid.
  • Inhibits α-Amylase, AChE, and BChE
  • Exhibits antioxidant activity, scavenging free radicals DPPH and ABTS
  • Suitable for research on diabetes and oxidative stress-associated diseases
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month
  • Aliquot and store stock solutions to prevent inactivation from repeated freeze-thaw cycles
  • For research use only
Catalog No. 50-003-60827 Supplier Medchemexpress LLC Supplier No. HY14900810MM/1ML
α-Amylase-IN-3 (Compound 4) is an inhibitor for α-Amylase, AChE, and BChE, with respective IC50 values of 18.04 μM, 21.04 μM, and 22.2 μM. It demonstrates antioxidant activity and is utilized in research concerning diabetes and oxidative stress-associated diseases. The product is supplied as a white to off-white solid.
  • Inhibits α-Amylase, AChE, and BChE
  • Exhibits antioxidant activity, scavenging free radicals DPPH and ABTS
  • Suitable for research on diabetes and oxidative stress-associated diseases
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month
  • Aliquot and store stock solutions to prevent inactivation from repeated freeze-thaw cycles
  • For research use only