Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Amylase-IN-3 | 93944-58-2 | 99.5% | 267.24 | 50 MG
SDP

Catalog No. 5000360899
Encompass_Preferred

α-Amylase-IN-3 is an inhibitor targeting α-Amylase, AChE, and BChE with IC50 values of 18.04 μM, 21.04 μM, and 22.2 μM, respectively. This compound also demonstrates significant antioxidant activity, making it a valuable tool for research into diabetes and various oxidative stress-associated diseases.

  • Inhibits α-Amylase, AChE, and BChE.
  • Exhibits antioxidant activity.
  • Useful in research for diabetes and oxidative stress-associated diseases.
  • High purity of 99.5%.
  • Molecular weight is 267.24.
  • Soluble in DMSO at 11.11 mg/mL.
  • Stable as a powder for up to 3 years at -20°C or 2 years at 4°C.
  • Stable in solvent for up to 6 months at -80°C or 1 month at -20°C.

Catalog No. 50-003-60899 Supplier Medchemexpress LLC Supplier No. HY14900850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Amylase-IN-3 is an inhibitor targeting α-Amylase, AChE, and BChE with IC50 values of 18.04 μM, 21.04 μM, and 22.2 μM, respectively. This compound also demonstrates significant antioxidant activity, making it a valuable tool for research into diabetes and various oxidative stress-associated diseases.

  • Inhibits α-Amylase, AChE, and BChE.
  • Exhibits antioxidant activity.
  • Useful in research for diabetes and oxidative stress-associated diseases.
  • High purity of 99.5%.
  • Molecular weight is 267.24.
  • Soluble in DMSO at 11.11 mg/mL.
  • Stable as a powder for up to 3 years at -20°C or 2 years at 4°C.
  • Stable in solvent for up to 6 months at -80°C or 1 month at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.