Learn More
Medchemexpress LLC α-amylase-IN-3 | 93944-58-2 | 99.5% | 267.24 g/mol | C15H9NO4 | 10 MG
SDP
Catalog No. 5000285376
Encompass_Preferred
α-Amylase-IN-3 is a small-molecule research reagent reported to inhibit α-amylase and acetylcholinesterase, and to exhibit antioxidant activity. It is supplied as a solid for laboratory research and is used in studies related to diabetes and oxidative stress-associated disease.
  • Non-competitive α-amylase inhibitor with reported IC50 ≈ 18.04 μM.
  • Acetylcholinesterase inhibitory activity with reported IC50s ≈ 21.04 μM and 22.2 μM.
  • Demonstrates antioxidant activity in standard radical-scavenging assays.
  • Supplied as a stable solid suitable for laboratory storage and handling.
  • High reported purity (≈99.49% by HPLC).
  • Molecular weight 267.24 g/mol; chemical formula C15H9NO4.
Catalog No. 50-002-85376 Supplier Medchemexpress LLC Supplier No. HY14900810MG
α-Amylase-IN-3 is a small-molecule research reagent reported to inhibit α-amylase and acetylcholinesterase, and to exhibit antioxidant activity. It is supplied as a solid for laboratory research and is used in studies related to diabetes and oxidative stress-associated disease.
  • Non-competitive α-amylase inhibitor with reported IC50 ≈ 18.04 μM.
  • Acetylcholinesterase inhibitory activity with reported IC50s ≈ 21.04 μM and 22.2 μM.
  • Demonstrates antioxidant activity in standard radical-scavenging assays.
  • Supplied as a stable solid suitable for laboratory storage and handling.
  • High reported purity (≈99.49% by HPLC).
  • Molecular weight 267.24 g/mol; chemical formula C15H9NO4.