Learn More
Medchemexpress LLC α-Amylase, Porcine Pancreatic | 9000-90-2 | 5 KU
SDP
Catalog No. 5000412246
Encompass_Preferred
α-Amylase, Porcine Pancreatic is a hydrolase enzyme isolated from porcine pancreatic. It catalyzes the hydrolysis of internal α-1, 4-glycosidic linkages in starch, producing glucose and maltose. This product is intended for life science research.
  • Used in the study of starch metabolism.
  • Shows binding activity toward N-linked oligosaccharides of glycoproteins.
  • Enhances the release of Dexamethasone from its γ-cyclodextrin complex.
Catalog No. 50-004-12246 Supplier Medchemexpress LLC Supplier No. HYB2193C5KU
α-Amylase, Porcine Pancreatic is a hydrolase enzyme isolated from porcine pancreatic. It catalyzes the hydrolysis of internal α-1, 4-glycosidic linkages in starch, producing glucose and maltose. This product is intended for life science research.
  • Used in the study of starch metabolism.
  • Shows binding activity toward N-linked oligosaccharides of glycoproteins.
  • Enhances the release of Dexamethasone from its γ-cyclodextrin complex.